RIE156324444 Rieke Metals, Inc.

ethyl 4-[4-(azetidinomethyl)phenyl]-4-oxobutyrate

  • MDL number MFCD03841604
  • Cas Number : 898757-19-2
  • Molecular Weight : 275.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-[4-(1-azetidinylmethyl)phenyl]-4-oxobutanoate
Inchi Code 1S/C16H21NO3/c1-2-20-16(19)9-8-15(18)14-6-4-13(5-7-14)12-17-10-3-11-17/h4-7H,2-3,8-12H2,1H3
Inchi Key KWZFYBJWTXOJAH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0405-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0405-00
Availability details for
Quantity
  • Availability
  • Add to Cart