RIE156324448 Rieke Metals, Inc.

ethyl 8-[4-(azetidinomethyl)phenyl]-8-oxooctanoate

  • MDL number MFCD07699561
  • Cas Number : 898757-31-8
  • Molecular Weight : 331.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0409-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0409-00
Availability details for
Quantity
  • Availability
  • Add to Cart