RIE156324463 Rieke Metals, Inc.

4'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-4-thiomethyl benzophenone

  • MDL number MFCD03841680
  • Cas Number : 898757-74-9
  • Molecular Weight : 383.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [4-(1,4-dioxa-8-azaspiro[4.5]dec-8-ylmethyl)phenyl][4-(methylsulfanyl)phenyl]methanone
Inchi Code 1S/C22H25NO3S/c1-27-20-8-6-19(7-9-20)21(24)18-4-2-17(3-5-18)16-23-12-10-22(11-13-23)25-14-15-26-22/h2-9H,10-16H2,1H3
Inchi Key ULWNEUGIWIZULC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0600-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0600-15
Availability details for
Quantity
  • Availability
  • Add to Cart