RIE156324464 Rieke Metals, Inc.

3-bromo-4'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]benzophenone

  • MDL number MFCD03841681
  • Cas Number : 898757-76-1
  • Molecular Weight : 416.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0600-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0600-18
Availability details for
Quantity
  • Availability
  • Add to Cart