RIE156324470 Rieke Metals, Inc.

2,3-dimethyl-4'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]benzophenone

  • MDL number MFCD03841687
  • Cas Number : 898757-88-5
  • Molecular Weight : 365.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0600-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0600-26
Availability details for
Quantity
  • Availability
  • Add to Cart