RIE156324477 Rieke Metals, Inc.

4-chloro-4'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-3-fluorobenzophenone

  • MDL number MFCD03841693
  • Cas Number : 898758-02-6
  • Molecular Weight : 389.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-chloro-3-fluorophenyl)[4-(1,4-dioxa-8-azaspiro[4.5]dec-8-ylmethyl)phenyl]methanone
Inchi Code 1S/C21H21ClFNO3/c22-18-6-5-17(13-19(18)23)20(25)16-3-1-15(2-4-16)14-24-9-7-21(8-10-24)26-11-12-27-21/h1-6,13H,7-12,14H2
Inchi Key PGWFXLCFMLCWES-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0600-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0600-33
Availability details for
Quantity
  • Availability
  • Add to Cart