RIE156324480 Rieke Metals, Inc.

4'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-2-fluorobenzophenone

  • MDL number MFCD03841696
  • Cas Number : 898758-08-2
  • Molecular Weight : 355.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0600-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0600-37
Availability details for
Quantity
  • Availability
  • Add to Cart