RIE156324483 Rieke Metals, Inc.

4'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-4-trifluoromethylbenzophenone

  • MDL number MFCD03841699
  • Cas Number : 898758-14-0
  • Molecular Weight : 405.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0600-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0600-45
Availability details for
Quantity
  • Availability
  • Add to Cart