RIE156324484 Rieke Metals, Inc.

4-bromo-4'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-2-fluorobenzophenone

  • MDL number MFCD03841700
  • Cas Number : 898758-16-2
  • Molecular Weight : 434.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0600-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0600-49
Availability details for
Quantity
  • Availability
  • Add to Cart