RIE156324495 Rieke Metals, Inc.

3,5-difluoro-4'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]benzophenone

  • MDL number MFCD03841711
  • Cas Number : 898758-49-1
  • Molecular Weight : 373.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-difluorophenyl)[4-(1,4-dioxa-8-azaspiro[4.5]dec-8-ylmethyl)phenyl]methanone
Inchi Code 1S/C21H21F2NO3/c22-18-11-17(12-19(23)13-18)20(25)16-3-1-15(2-4-16)14-24-7-5-21(6-8-24)26-9-10-27-21/h1-4,11-13H,5-10,14H2
Inchi Key YCQRHYYERBIJNL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0600-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0600-60
Availability details for
Quantity
  • Availability
  • Add to Cart