RIE156324496 Rieke Metals, Inc.

4'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-3,4,5-trifluorobenzophenone

  • MDL number MFCD03841712
  • Cas Number : 898758-52-6
  • Molecular Weight : 391.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0600-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0600-61
Availability details for
Quantity
  • Availability
  • Add to Cart