RIE156324502 Rieke Metals, Inc.

ethyl 5-[4-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]phenyl]-5-oxovalerate

  • MDL number MFCD03841718
  • Cas Number : 898782-07-5
  • Molecular Weight : 375.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-[4-(1,4-dioxa-8-azaspiro[4.5]dec-8-ylmethyl)phenyl]-5-oxopentanoate
Inchi Code 1S/C21H29NO5/c1-2-25-20(24)5-3-4-19(23)18-8-6-17(7-9-18)16-22-12-10-21(11-13-22)26-14-15-27-21/h6-9H,2-5,10-16H2,1H3
Inchi Key HVAJCABNCJCUGO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0606-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0606-00
Availability details for
Quantity
  • Availability
  • Add to Cart