RIE156324505 Rieke Metals, Inc.

ethyl 8-[4-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]phenyl]-8-oxooctanoate

  • MDL number MFCD03841721
  • Cas Number : 898782-16-6
  • Molecular Weight : 417.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0609-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0609-00
Availability details for
Quantity
  • Availability
  • Add to Cart