RIE156324509 Rieke Metals, Inc.

4-methyl-4'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03841725
  • Cas Number : 898782-28-0
  • Molecular Weight : 311.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-methylphenyl)[4-(4-thiomorpholinylmethyl)phenyl]methanone
Inchi Code 1S/C19H21NOS/c1-15-2-6-17(7-3-15)19(21)18-8-4-16(5-9-18)14-20-10-12-22-13-11-20/h2-9H,10-14H2,1H3
Inchi Key IHGCJZZOIOYLLY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0700-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-04
Availability details for
Quantity
  • Availability
  • Add to Cart