RIE156324510 Rieke Metals, Inc.

2-methoxy-4'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03841726
  • Cas Number : 898782-31-5
  • Molecular Weight : 327.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0700-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-05
Availability details for
Quantity
  • Availability
  • Add to Cart