RIE156324515 Rieke Metals, Inc.

4-cyano-4'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03841731
  • Cas Number : 898782-46-2
  • Molecular Weight : 322.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0700-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-10
Availability details for
Quantity
  • Availability
  • Add to Cart