RIE156324520 Rieke Metals, Inc.

4-thiomethyl-4'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03841736
  • Cas Number : 898782-57-5
  • Molecular Weight : 343.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0700-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-15
Availability details for
Quantity
  • Availability
  • Add to Cart