RIE156324523 Rieke Metals, Inc.

3-chloro-4'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03841739
  • Cas Number : 898782-63-3
  • Molecular Weight : 331.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0700-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-20
Availability details for
Quantity
  • Availability
  • Add to Cart