RIE156324531 Rieke Metals, Inc.

3,4-dimethyl-4'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03841747
  • Cas Number : 898782-79-1
  • Molecular Weight : 325.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0700-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-30
Availability details for
Quantity
  • Availability
  • Add to Cart