RIE156324533 Rieke Metals, Inc.

4-bromo-3-fluoro-4'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03841749
  • Cas Number : 898782-83-7
  • Molecular Weight : 394.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromo-3-fluorophenyl)[4-(4-thiomorpholinylmethyl)phenyl]methanone
Inchi Code 1S/C18H17BrFNOS/c19-16-6-5-15(11-17(16)20)18(22)14-3-1-13(2-4-14)12-21-7-9-23-10-8-21/h1-6,11H,7-10,12H2
Inchi Key QQIQBARQVCHCLE-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0700-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-32
Availability details for
Quantity
  • Availability
  • Add to Cart