RIE156324534 Rieke Metals, Inc.

4-chloro-3-fluoro-4'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03841750
  • Cas Number : 898782-85-9
  • Molecular Weight : 349.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0700-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-33
Availability details for
Quantity
  • Availability
  • Add to Cart