RIE156324536 Rieke Metals, Inc.

2-chloro-4'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03841752
  • Cas Number : 898782-90-6
  • Molecular Weight : 331.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0700-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-36
Availability details for
Quantity
  • Availability
  • Add to Cart