RIE156324539 Rieke Metals, Inc.

4'-thiomorpholinomethyl-3-trifluoromethylbenzophenone

  • MDL number MFCD03841755
  • Cas Number : 898782-98-4
  • Molecular Weight : 365.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [4-(4-thiomorpholinylmethyl)phenyl][3-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C19H18F3NOS/c20-19(21,22)17-3-1-2-16(12-17)18(24)15-6-4-14(5-7-15)13-23-8-10-25-11-9-23/h1-7,12H,8-11,13H2
Inchi Key JXZDRNUVYSTIPU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0700-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-44
Availability details for
Quantity
  • Availability
  • Add to Cart