RIE156324540 Rieke Metals, Inc.

4-thiomorpholinomethyl-4'-trifluoromethylbenzophenone

  • MDL number MFCD03841756
  • Cas Number : 898783-00-1
  • Molecular Weight : 365.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0700-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-45
Availability details for
Quantity
  • Availability
  • Add to Cart