RIE156324542 Rieke Metals, Inc.

2-chloro-4-fluoro-4'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03841758
  • Cas Number : 898783-04-5
  • Molecular Weight : 349.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chloro-4-fluorophenyl)[4-(4-thiomorpholinylmethyl)phenyl]methanone
Inchi Code 1S/C18H17ClFNOS/c19-17-11-15(20)5-6-16(17)18(22)14-3-1-13(2-4-14)12-21-7-9-23-10-8-21/h1-6,11H,7-10,12H2
Inchi Key GEFBZFKRYIRXFT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0700-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-50
Availability details for
Quantity
  • Availability
  • Add to Cart