RIE156324543 Rieke Metals, Inc.

3-chloro-5-fluoro-4'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03841759
  • Cas Number : 898783-06-7
  • Molecular Weight : 349.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chloro-5-fluorophenyl)[4-(4-thiomorpholinylmethyl)phenyl]methanone
Inchi Code 1S/C18H17ClFNOS/c19-16-9-15(10-17(20)11-16)18(22)14-3-1-13(2-4-14)12-21-5-7-23-8-6-21/h1-4,9-11H,5-8,12H2
Inchi Key IJMJBVBSFMQJEU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0700-51

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-51
Availability details for
Quantity
  • Availability
  • Add to Cart