RIE156324544 Rieke Metals, Inc.

4-chloro-2-fluoro-4'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03841760
  • Cas Number : 898783-08-9
  • Molecular Weight : 349.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-chloro-2-fluorophenyl)[4-(4-thiomorpholinylmethyl)phenyl]methanone
Inchi Code 1S/C18H17ClFNOS/c19-15-5-6-16(17(20)11-15)18(22)14-3-1-13(2-4-14)12-21-7-9-23-10-8-21/h1-6,11H,7-10,12H2
Inchi Key YEOPXTNGVIRQSQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0700-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-52
Availability details for
Quantity
  • Availability
  • Add to Cart