RIE156324546 Rieke Metals, Inc.

2,4-dichloro-4'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03841762
  • Cas Number : 898783-12-5
  • Molecular Weight : 366.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0700-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-54
Availability details for
Quantity
  • Availability
  • Add to Cart