RIE156324547 Rieke Metals, Inc.

2,5-dichloro-4'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03841763
  • Cas Number : 898783-14-7
  • Molecular Weight : 366.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0700-55

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-55
Availability details for
Quantity
  • Availability
  • Add to Cart