RIE156324553 Rieke Metals, Inc.

4'-thiomorpholinomethyl-3,4,5-trifluorobenzophenone

  • MDL number MFCD03841769
  • Cas Number : 898783-26-1
  • Molecular Weight : 351.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0700-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0700-61
Availability details for
Quantity
  • Availability
  • Add to Cart