RIE156324561 Rieke Metals, Inc.

ethyl 7-oxo-7-[4-(thiomorpholinomethyl)phenyl]heptanoate

  • MDL number MFCD03841777
  • Cas Number : 898783-40-9
  • Molecular Weight : 363.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0708-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0708-00
Availability details for
Quantity
  • Availability
  • Add to Cart