RIE156324563 Rieke Metals, Inc.

4-(4-methylpiperazinomethyl)benzophenone

  • MDL number MFCD03841779
  • Cas Number : 898783-42-1
  • Molecular Weight : 294.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name {4-[(4-methyl-1-piperazinyl)methyl]phenyl}(phenyl)methanone
Inchi Code 1S/C19H22N2O/c1-20-11-13-21(14-12-20)15-16-7-9-18(10-8-16)19(22)17-5-3-2-4-6-17/h2-10H,11-15H2,1H3
Inchi Key YRSRJTSAWRCXIO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0800-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0800-01
Availability details for
Quantity
  • Availability
  • Add to Cart