RIE156324564 Rieke Metals, Inc.

2-methyl-4'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03841780
  • Cas Number : 898783-43-2
  • Molecular Weight : 308.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methylphenyl){4-[(4-methyl-1-piperazinyl)methyl]phenyl}methanone
Inchi Code 1S/C20H24N2O/c1-16-5-3-4-6-19(16)20(23)18-9-7-17(8-10-18)15-22-13-11-21(2)12-14-22/h3-10H,11-15H2,1-2H3
Inchi Key QHUGMTNDWGBYSA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0800-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0800-02
Availability details for
Quantity
  • Availability
  • Add to Cart