RIE156324580 Rieke Metals, Inc.

3-chloro-4'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03841796
  • Cas Number : 898783-61-4
  • Molecular Weight : 328.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chlorophenyl){4-[(4-methyl-1-piperazinyl)methyl]phenyl}methanone
Inchi Code 1S/C19H21ClN2O/c1-21-9-11-22(12-10-21)14-15-5-7-16(8-6-15)19(23)17-3-2-4-18(20)13-17/h2-8,13H,9-12,14H2,1H3
Inchi Key QCSKJBREPSXYHS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0800-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0800-20
Availability details for
Quantity
  • Availability
  • Add to Cart