RIE156324584 Rieke Metals, Inc.

2,3-dimethyl-4'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03841800
  • Cas Number : 898783-69-2
  • Molecular Weight : 322.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,3-dimethylphenyl){4-[(4-methyl-1-piperazinyl)methyl]phenyl}methanone
Inchi Code 1S/C21H26N2O/c1-16-5-4-6-20(17(16)2)21(24)19-9-7-18(8-10-19)15-23-13-11-22(3)12-14-23/h4-10H,11-15H2,1-3H3
Inchi Key HICHZMKYLBHYOT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0800-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0800-26
Availability details for
Quantity
  • Availability
  • Add to Cart