RIE156324593 Rieke Metals, Inc.

2-chloro-4'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03841809
  • Cas Number : 898783-87-4
  • Molecular Weight : 328.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chlorophenyl){4-[(4-methyl-1-piperazinyl)methyl]phenyl}methanone
Inchi Code 1S/C19H21ClN2O/c1-21-10-12-22(13-11-21)14-15-6-8-16(9-7-15)19(23)17-4-2-3-5-18(17)20/h2-9H,10-14H2,1H3
Inchi Key FFIONIBFQWFYDO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0800-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0800-36
Availability details for
Quantity
  • Availability
  • Add to Cart