RIE156324595 Rieke Metals, Inc.

4'-(4-methylpiperazinomethyl)-2-trifluoromethylbenzophenone

  • MDL number MFCD03841811
  • Cas Number : 898783-91-0
  • Molecular Weight : 362.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0800-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0800-43
Availability details for
Quantity
  • Availability
  • Add to Cart