RIE156324596 Rieke Metals, Inc.

4'-(4-methylpiperazinomethyl)-3-trifluoromethylbenzophenone

  • MDL number MFCD03841812
  • Cas Number : 898783-93-2
  • Molecular Weight : 362.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0800-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0800-44
Availability details for
Quantity
  • Availability
  • Add to Cart