RIE156324598 Rieke Metals, Inc.

4-bromo-2-fluoro-4'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03841814
  • Cas Number : 898783-97-6
  • Molecular Weight : 391.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0800-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0800-49
Availability details for
Quantity
  • Availability
  • Add to Cart