RIE156324601 Rieke Metals, Inc.

4-chloro-2-fluoro-4'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03841817
  • Cas Number : 898763-15-0
  • Molecular Weight : 346.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0800-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0800-52
Availability details for
Quantity
  • Availability
  • Add to Cart