RIE156324606 Rieke Metals, Inc.

3,5-dichloro-4'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03841822
  • Cas Number : 898763-30-9
  • Molecular Weight : 363.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0800-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0800-57
Availability details for
Quantity
  • Availability
  • Add to Cart