RIE156324609 Rieke Metals, Inc.

3,5-difluoro-4'-(4-methylpiperazinomethyl) benzophenone

  • MDL number MFCD03841825
  • Cas Number : 898763-39-8
  • Molecular Weight : 330.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0800-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0800-60
Availability details for
Quantity
  • Availability
  • Add to Cart