RIE156324610 Rieke Metals, Inc.

4'-(4-methylpiperazinomethyl)-3,4,5-trifluorobenzophenone

  • MDL number MFCD03841826
  • Cas Number : 898763-42-3
  • Molecular Weight : 348.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0800-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0800-61
Availability details for
Quantity
  • Availability
  • Add to Cart