RIE156324611 Rieke Metals, Inc.

cyclopropyl 4-(4-methylpiperazinomethyl)phenyl ketone

  • MDL number MFCD03841827
  • Cas Number : 898763-45-6
  • Molecular Weight : 258.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0801-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0801-00
Availability details for
Quantity
  • Availability
  • Add to Cart