RIE156324612 Rieke Metals, Inc.

cyclobutyl 4-(4-methylpiperazinomethyl)phenyl ketone

  • MDL number MFCD03841828
  • Cas Number : 898763-48-9
  • Molecular Weight : 272.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0802-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0802-00
Availability details for
Quantity
  • Availability
  • Add to Cart