RIE156324615 Rieke Metals, Inc.

ethyl 4-[4-(4-methylpiperazinomethyl)phenyl]-4-oxobutyrate

  • MDL number MFCD03841831
  • Cas Number : 898763-57-0
  • Molecular Weight : 318.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-{4-[(4-methyl-1-piperazinyl)methyl]phenyl}-4-oxobutanoate
Inchi Code 1S/C18H26N2O3/c1-3-23-18(22)9-8-17(21)16-6-4-15(5-7-16)14-20-12-10-19(2)11-13-20/h4-7H,3,8-14H2,1-2H3
Inchi Key GVHCDIZODDPFQI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0805-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0805-00
Availability details for
Quantity
  • Availability
  • Add to Cart