RIE156324618 Rieke Metals, Inc.

ethyl 7-[4-(4-methylpiperazinomethyl)phenyl]-7-oxoheptanoate

  • MDL number MFCD03841834
  • Cas Number : 898763-65-0
  • Molecular Weight : 360.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0808-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0808-00
Availability details for
Quantity
  • Availability
  • Add to Cart