RIE156324619 Rieke Metals, Inc.

ethyl 8-[4-(4-methylpiperazinomethyl)phenyl]-8-oxooctanoate

  • MDL number MFCD03841835
  • Cas Number : 898763-67-2
  • Molecular Weight : 374.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 8-{4-[(4-methyl-1-piperazinyl)methyl]phenyl}-8-oxooctanoate
Inchi Code 1S/C22H34N2O3/c1-3-27-22(26)9-7-5-4-6-8-21(25)20-12-10-19(11-13-20)18-24-16-14-23(2)15-17-24/h10-13H,3-9,14-18H2,1-2H3
Inchi Key AEAJQQIOFVZKEM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0809-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0809-00
Availability details for
Quantity
  • Availability
  • Add to Cart