RIE156324630 Rieke Metals, Inc.

2-carboethoxy-4'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03841846
  • Cas Number : 898763-89-8
  • Molecular Weight : 335.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-[4-(2,5-dihydro-1H-pyrrol-1-ylmethyl)benzoyl]benzoate
Inchi Code 1S/C21H21NO3/c1-2-25-21(24)19-8-4-3-7-18(19)20(23)17-11-9-16(10-12-17)15-22-13-5-6-14-22/h3-12H,2,13-15H2,1H3
Inchi Key TYZPSASIHVVMPD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6124-0900-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0900-11
Availability details for
Quantity
  • Availability
  • Add to Cart