RIE156324632 Rieke Metals, Inc.

4-carboethoxy-4'-(3-pyrrolinomethyl) benzophenone

  • MDL number MFCD03841848
  • Cas Number : 898763-93-4
  • Molecular Weight : 335.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0900-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0900-13
Availability details for
Quantity
  • Availability
  • Add to Cart